Learn More
ACROBiosystems Human CXCR5 20ug
Catalog No. 502535185
Encompass_Preferred
Human CXCR5 Protein FlagHis Tag (CX5-H52D6) is expressed from human 293 cells (HEK293). It contains AA Asn 2 - Phe 372 (Accession # P32302-1).
Catalog No. 50-253-5185 Supplier ACROBiosystems Supplier No. CX5H52D620UG
Human CXCR5 Protein FlagHis Tag (CX5-H52D6) is expressed from human 293 cells (HEK293). It contains AA Asn 2 - Phe 372 (Accession # P32302-1).