Learn More
Sino Biological Recombinant Human CXCR6 Protein (VLP, Full-Length, Flag & His-Strep Tag)
Catalog No. 502993119
Encompass_Preferred
A DNA sequence encoding the Human CXCR6 (O00574) (Met1-Leu342) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.
Catalog No. 50-299-3119 Supplier Sino Biological Supplier No. 12504H82H20
A DNA sequence encoding the Human CXCR6 (O00574) (Met1-Leu342) was expressed, with a Flag tag at the N-terminus, a polyhistidine tag and a strep tag at the C-terminus.