Learn More
BIOHIPPO Human CXCR6 Microsample
SDP
Catalog No. 5000313835
Encompass
Human CXCR6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CXCR6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 051 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CXCR6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CXCR6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CXCR6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13835 Supplier BIOHIPPO Supplier No. BHE15211616
Human CXCR6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CXCR6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 051 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CXCR6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CXCR6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CXCR6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color