Learn More
Proteintech Group Inc Human CYB5R3 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0006996T
Encompass_Preferred
CYB5R3, also named as DIA1 and B5R, belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in red blood cells. There are two types of methemoglobinemia CYB5R3-related. In type 1, the defect affects the soluble form of the enzyme, is restricted to red blood cells, and causes well-tolerated methemoglobinemia. In type 2, the defect affects both the soluble and microsomal forms of the enzyme and is thus generalized, affecting red cells, leukocytes and all body tissues. Type 2 methemoglobinemia is associated with mental deficiency and other neurologic symptoms. The ELISA kit is suitable for testing serum, plasma, and cell lysates.
Catalog No. 50-197-4035