Learn More
Biomatik Corporation Human Cyclin A1 (CCNA1) ELISA Kit
Supplier:  Biomatik Corporation EKU03528
Encompass_Preferred
Human Cyclin A1 (CCNA1) ELISA Kit for detecting CCNA1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P78396. Gene ID: 8900. Target Synonyms: CCN-A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4741