Learn More
Biomatik Corporation Human Cyclin D3 (CCND3) ELISA Kit
Supplier:  Biomatik Corporation EKU10100
Encompass_Preferred
Human Cyclin D3 (CCND3) ELISA Kit for detecting CCND3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30281. Gene ID: 896. Target Synonyms: CCN-D3; G1/S-Specific Cyclin-D3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8371