Learn More
Biomatik Corporation Human Cyclin Dependent Kinase Inhibitor 1A (CDKN1A) ELISA Kit
Supplier:  Biomatik Corporation EKU10764
Encompass_Preferred
Human Cyclin Dependent Kinase Inhibitor 1A (CDKN1A) ELISA Kit for detecting CDKN1A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P38936. Gene ID: 1026. Target Synonyms: WAF1; p21; CIP1; p21WAF1; CAP20; CDKN1; MDA-6; SDI1; p21CIP1; CDK-Interacting Protein 1; Melanoma differentiation-associated protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4423