Learn More
Biomatik Corporation Human Cyclin Dependent Kinase Inhibitor 1B (CDKN1B) ELISA Kit
Supplier:  Biomatik Corporation EKU11761
Encompass_Preferred
Human Cyclin Dependent Kinase Inhibitor 1B (CDKN1B) ELISA Kit for detecting CDKN1B in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.236ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46527. Gene ID: 1027. Target Synonyms: CDKN4; KIP1; P27KIP1; p27; Cyclin-dependent kinase inhibitor p27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5633