Learn More
Biomatik Corporation Human Cyclin Dependent Kinase 2 (CDK2) ELISA Kit
Supplier:  Biomatik Corporation EKU03536
Encompass_Preferred
Human Cyclin Dependent Kinase 2 (CDK2) ELISA Kit for detecting CDK2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24941. Gene ID: 1017. Target Synonyms: CDKN2; p33 (CDK2) ; Cell division protein kinase 2; p33 protein kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9026