Learn More
Biomatik Corporation Human Cyclin Dependent Kinase Inhibitor 3 (CDKN3) ELISA Kit
Supplier:  Biomatik Corporation EKU03547
Encompass_Preferred
Human Cyclin Dependent Kinase Inhibitor 3 (CDKN3) ELISA Kit for detecting CDKN3 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16667. Gene ID: 1033. Target Synonyms: CDI1; CIP2; KAP1; CDK2-Associated Dual Specificity Phosphatase; Kinase Associated Phosphatase; Cyclin-dependent kinase interactor 1; Cyclin-dependent kinase-interacting 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9023