Learn More
ACROBiosystems ACROBiosystems Human / Cynomolgus / Rhesus macaque ROR1 (39-151, Ig-like domain) Protein, His Tag
Supplier:  ACROBiosystems RO1H52211MG
Encompass_Preferred
Recombinant Protein;1MG;Human / Cynomolgus / Rhesus macaque ROR1 (39-151, Ig-like domain), His Tag (RO1-H5221) is expressed from human 293 cells (HEK293). It contains AA Glu 39 - Gly 151 (Accession # Q01973-1). In the region Gln 30 - Glu 403, the AA sequence of Human, Cynomolgus and Rhesus macaque ROR1 are homologus.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 13.7 kDa. The protein migrates as 25-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human / Cynomolgus / Rhesus macaque ROR1 (39-151, Ig-like domain), His Tag (Cat. No. RO1-H5221) at 2 μg/mL (100 μL/well) can bind Anti-Human ROR1 MAb with a linear range of 1-31 ng/mL (QC tested).
Catalog No. 50-210-6750