Learn More
Sino Biological VEGF165 Protein, Human, Recombinant (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 11066H27HB100
Encompass_Preferred
A DNA sequence encoding the human, Cynomolgus VEGF165 (P15692-4) (Ala27-Arg191) was expressed with a C-terminal AVI tag followed by a polyhistidine tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4895