Learn More
Sino Biological Human, Cynomolgus VEGFA MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11066MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human / Cynomolgus VEGF165/VEGFA (rh VEGF165/VEGFA; Catalog#11066-HNAH; P15692-4; Met1-Arg191). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-0926