Learn More
Sino Biological Human, Cynomolgus VEGFA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40010T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human / Cynomolgus VEGF165/VEGFA (rh VEGF165/VEGFA; Catalog#11066-HNAB; P15692-4; Met1-Arg191). VEGF165/VEGFA specific IgG was purified by Human / Cynomolgus VEGF165/VEGFA affinity chromatography.
Catalog No. 50-237-4077