Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CYP11B1 (aa 156-242) Control Fragment Recombinant Protein

Catalog No. RP104168
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104168 Supplier Invitrogen™ Supplier No. RP104168
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63290. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The steroid 11beta-hydroxylase gene, also designated Cyp11b-1, is a marker for the functional differentiation of cells in the zonae fasciculata reticularis. The deduced protein CYP11B1 consists of 466 amino acids containing a secretory signal, epidermal growth factor-like repeats, and a proteolytically inactive cathepsin B-related sequence. A related protein, human aldosterone synthase (CYP11B2), is involved in substrate recognition and conversion, with a functionally significant residue 112 in the N-terminal region of human CYP11B2. The inherited disorder glucocorticoid-remediable aldosteronism is caused by a chimeric gene duplication between the CYP11B1 and CYP11B2 genes. This disorder is characterized by hyperaldosteronism and high levels of 18-hydroxycortisol and 18-oxocortisol, which are under ACTH control.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15538
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 1584
Name Human CYP11B1 (aa 156-242) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 11-beta-hydroxylase; AA389061; aldosterone synthase; Cp11ba; CPN1; CYP11B; CYP11B1; Cyp11b-1; CYPXIB1; Cytochrome P450 11B1, mitochondrial; cytochrome P450 family 11 subfamily B member 1; cytochrome p450 XIB1; cytochrome P450(11 beta)-DS; cytochrome P450, family 11, subfamily b, polypeptide 1; cytochrome P450, subfamily 11B, polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; Cytochrome P450, subfamily XIB, polypeptide 1 (steroid 11-beta-hydroxylase); cytochrome P450C11; cytochrome P-450c11; DKFZp686B05283; FHI; FLJ36771; P450(11 beta)-DS; P450C 11; P450C11; RATCP11BA; S11BH; steroid 11-beta-hydroxylase; steroid 11-beta-hydroxylase, CYP11B1; steroid 11-beta-monooxygenase; steroid-11-beta-hydroxylase
Common Name CYP11B1
Gene Symbol CYP11B1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RFLPMVDAVARDFSQALKKKVLQNARGSLTLDVQPSIFHYTIEASNLALFGERLGLVGHSPSSASLNFLHALEVMFKSTVQLMFMPR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.