Learn More
Discovery Life Sciences HUMAN CYP1A1+REDUCTASE 0.5NMOL
SDP
Catalog No. NC9758150
Encompass
Human CYP1A1 + P450 Reductase SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP1A1 which is expressed from human CYP1A1 cDNA using a baculovirus expression system
Catalog No. NC9758150 Supplier Discovery Life Sciences Supplier No. 456211
Human CYP1A1 + P450 Reductase SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP1A1 which is expressed from human CYP1A1 cDNA using a baculovirus expression system