Learn More
Discovery Life Sciences HUMAN CYP1A2+REDUCTASE 0.5NMOL
SDP
Catalog No. NC9496064
Encompass
Human CYP1A2 + P450 Reductase SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP1A2 which is expressed from human CYP1A2 cDNA using a baculovirus expression system
Catalog No. NC9496064 Supplier Discovery Life Sciences Supplier No. 456203
Human CYP1A2 + P450 Reductase SUPERSOMES 0.5 nmole cytochrome P450 in 0.5mL This activity is catalyzed by CYP1A2 which is expressed from human CYP1A2 cDNA using a baculovirus expression system