Learn More
BIOHIPPO Human CYP2A6 Microsample
SDP
Catalog No. 5000313860
Encompass
Human CYP2A6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CYP2A6 in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CYP2A6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CYP2A6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CYP2A6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13860 Supplier BIOHIPPO Supplier No. BHE15210605
Human CYP2A6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CYP2A6 in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CYP2A6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CYP2A6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CYP2A6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color