Learn More
BIOHIPPO Human CYR61 Microsample
SDP
Catalog No. 5000315604
Encompass
Human CYR61 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CYR61 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CYR61 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CYR61 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CYR61 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15604 Supplier BIOHIPPO Supplier No. BHE15211724
Human CYR61 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CYR61 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CYR61 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CYR61 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CYR61 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color