Learn More
Advanced ImmunoChemical, Inc. Human Cystatin C, 0.05mg
Supplier:  Advanced ImmunoChemical, Inc. 8CYC0.05MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Cystatis C, human. Source:Pooled human serum.Material was tested and found negative for HBsAg, HCV, HIV-1and 2 antibodies, HCV antibodies, and syphilis.Applications:Calibrator or standard in immunoassaysAnalysis:Purity > 95 % (SDS-PAGE)Immunological activity:Immunoreactivity is checked using monoclonal antibodies specific to cystatin CPurification:Affinity and size-exclusion chromatographyPresentation:Lyophilized from solution in 10 mM Tris-HCl, pH 7.5, 150 mM NaClProduct is recommended to reconstitute with deionized waterStorage Conditions:Before and after reconstitution at -20 °C (between -15 and -30 °C allowed)Other information:Avoid repeated freezing and thawing.It is recommended to aliquot the product after reconstitution.Material safety note:This product is sold for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. 50-153-9856