Learn More
Sino Biological Recombinant Mouse IL-16 Protein (His Tag)
Supplier:  Sino Biological 51303M07E50
Encompass_Preferred
A DNA sequence encoding the mouse IL16 (NP_034681.2) (Ser1205-Ser1322) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-6816