Learn More
Sino Biological Recombinant Human Nectin-2 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10005H49HB20
Encompass_Preferred
A DNA sequence encoding the human NECTIN2 (NP_002847.1)(Met1-Leu360) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-224-6815