Learn More
Biomatik Corporation Human Cystatin C (Cys-C) CLIA Kit
Supplier:  Biomatik Corporation EKU08924
Encompass_Preferred
Human Cystatin C (Cys-C) CLIA Kit for detecting Cys-C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.12ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01034. Gene ID: 1471. Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4883