Learn More
Raybiotech Inc Human Cystatin C ELISA Kit
Supplier:  Raybiotech Inc ELHCYSTATINC1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 300 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Species Reactivity Human Specificity Detects human Cystatin C in ELISAs. In sandwich ELISA assay, no cross reactivity with recombinant human Cystatin A, B, D, E/M, F, S, SA, SN, X/Z/P, or recombinant mouse Cystatin C is observed.
Catalog No. 50-194-9172