Learn More
Raybiotech Inc Human Cystatin F ELISA Kit
Supplier:  Raybiotech Inc ELHCYSTATINF1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cystatin F.
Catalog No. 50-194-9175