Learn More
Sino Biological Recombinant Human I-309/CCL1 Protein (hFc Tag)
Supplier:  Sino Biological 10057H01H100
Encompass_Preferred
A DNA sequence encoding the mature form of human CCL1 (NP_002972.1) (Lys 24-Lys 96) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-224-6958