Learn More
American Research Products Inc Human Cystatin SN CST1 ELISA
Catalog No. 5000328146
Encompass
Human Cystatin-SN (CST1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates urine tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28146 Supplier American Research Products Inc Supplier No. CSBEL006088HU
Human Cystatin-SN (CST1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates urine tissue homogenates cell lysates Method Sandwich