Learn More
Raybiotech Inc Human Cystatin SN ELISA Kit
Supplier:  Raybiotech Inc ELHCYSTATINSN1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Cystatin SN. Other species not yet determined.
Catalog No. 50-194-9176