Learn More
Biomatik Corporation Human Cysteine Rich Secretory Protein 2 (CRISP2) ELISA Kit
Supplier:  Biomatik Corporation EKU03575
Encompass_Preferred
Human Cysteine Rich Secretory Protein 2 (CRISP2) ELISA Kit for detecting CRISP2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16562. Gene ID: 7180. Target Synonyms: TSP1; CRISP-2; GAPDL5; TPX1; CT36; Testis Specific Protein 1; Cancer/Testis Antigen 36; Testis-specific protein TPX-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4999