Learn More
Biomatik Corporation Human Cysteine Sulfinic Acid Decarboxylase (CSAD) ELISA Kit
Supplier:  Biomatik Corporation EKU12059
Encompass_Preferred
Human Cysteine Sulfinic Acid Decarboxylase (CSAD) ELISA Kit for detecting CSAD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y600. Gene ID: 51380. Target Synonyms: PCAP; CSD; P-Selectin Cytoplasmic Tail-Associated Protein; Sulfinoalanine decarboxylase; Cysteine-sulfinate decarboxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6950