Learn More
Biomatik Corporation Human Cystin 1 (CYS1) ELISA Kit
Supplier:  Biomatik Corporation EKU09266
Encompass_Preferred
Human Cystin 1 (CYS1) ELISA Kit for detecting CYS1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q717R9. Gene ID: 192668. Target Synonyms: Ck; cpk; Cilia-Associated Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5203