Learn More
Biomatik Corporation Human Cytochrome b (COB) ELISA Kit
Supplier:  Biomatik Corporation EKU09357
Encompass_Preferred
Human Cytochrome b (COB) ELISA Kit for detecting COB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00156. Gene ID: 4519. Target Synonyms: MT-CYB; MTCYB; CYTB; UQCR3; Mitochondrially Encoded Cytochrome b; Cytochrome b-c1 complex subunit 3; Ubiquinol-cytochrome-c reductase complex cytochrome b subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6824