Learn More
Biomatik Corporation Human Cytochrome C Oxidase Subunit IV Isoform 1 (COX4I1) ELISA Kit
Supplier:  Biomatik Corporation EKU03585
Encompass_Preferred
Human Cytochrome C Oxidase Subunit IV Isoform 1 (COX4I1) ELISA Kit for detecting COX4I1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13073. Gene ID: 1327. Target Synonyms: COX4; Cytochrome c oxidase polypeptide IV; Cytochrome c oxidase subunit 4 isoform 1, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5098