Learn More
Biomatik Corporation Human Cytochrome P450 2B6 (CYP2B6) ELISA Kit
Supplier:  Biomatik Corporation EKU09155
Encompass_Preferred
Human Cytochrome P450 2B6 (CYP2B6) ELISA Kit for detecting CYP2B6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20813. Gene ID: 1555. Target Synonyms: CYP2B; CPB6; CYPIIB6; Cytochrome P450,Subfamily IIB (Phenobarbital-Inducible) Polypeptide 6; 1,4-cineole 2-exo-monooxygenase; Cytochrome P450 IIB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6766