Learn More
Biomatik Corporation Human Cytochrome P450 2C19 (CYP2C19) ELISA Kit
Supplier:  Biomatik Corporation EKU10782
Encompass_Preferred
Human Cytochrome P450 2C19 (CYP2C19) ELISA Kit for detecting CYP2C19 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33261. Gene ID: 1557. Target Synonyms: CPCJ; CYP2C; CYPIIC17; CYPIIC19; P450C2C; Cytochrome P450-11A; Cytochrome P450-254C; Mephenytoin 4-hydroxylase; limonene 6-monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8291