Learn More
Biomatik Corporation Human Cytochrome P450 2D6 (CYP2D6) ELISA Kit
Supplier:  Biomatik Corporation EKU09157
Encompass_Preferred
Human Cytochrome P450 2D6 (CYP2D6) ELISA Kit for detecting CYP2D6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10635. Gene ID: 1565. Target Synonyms: CYP2D; P450C2D; CPD6; CYP2DL1; P450-DB1; Cytochrome P450 Family 2 Subfamily D Polypeptide 6; Cytochrome P450-DB1; Debrisoquine 4-hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8559