Learn More
Biomatik Corporation Human Cytochrome P450 3A7 (CYP3A7) ELISA Kit
Supplier:  Biomatik Corporation EKU09353
Encompass_Preferred
Human Cytochrome P450 3A7 (CYP3A7) ELISA Kit for detecting CYP3A7 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.27ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24462. Gene ID: 100861540. Target Synonyms: CP37; P450-HFLA; Cytochrome P450,Family 3,Subfamily A,Polypeptide 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7310