Learn More
Biomatik Corporation Human Cytochrome P450 7B1 (CYP7B1) ELISA Kit
Supplier:  Biomatik Corporation EKU12025
Encompass_Preferred
Human Cytochrome P450 7B1 (CYP7B1) ELISA Kit for detecting CYP7B1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75881. Gene ID: 9420. Target Synonyms: CP7B; Oxysterol 7 Alpha-Hydroxylase; Spastic Paraplegia 5A (Autosomal Recessive) ; 25-hydroxycholesterol 7-alpha-hydroxylase; Oxysterol 7-alpha-hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8050