Learn More
Biomatik Corporation Human Cytokeratin 12 (CK12) ELISA Kit
Supplier:  Biomatik Corporation EKU05462
Encompass_Preferred
Human Cytokeratin 12 (CK12) ELISA Kit for detecting CK12 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99456. Gene ID: 3859. Target Synonyms: KRT12; K12; Keratin 12; Meesmann Corneal Dystrophy; Keratin, type I cytoskeletal 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7047