Learn More
Abcam Human Cytokeratin 18 Fragment ELISA Kit (CK18-30), 1 x 96 Tests
Catalog No. 502656978
Encompass_Preferred
Human Cytokeratin 18 Fragment ELISA Kit (CK18-30) is a single-wash 90-min Simplestep used to quantify Human Cytokeratin 18 Fragment (CK18-30) with a sensitivity of 22.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6978 Supplier Abcam Supplier No. AB2545151X96TESTS
Human Cytokeratin 18 Fragment ELISA Kit (CK18-30) is a single-wash 90-min Simplestep used to quantify Human Cytokeratin 18 Fragment (CK18-30) with a sensitivity of 22.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement