Learn More
Invitrogen™ Human Cytokeratin 8/KRT8 ELISA Kit
Description
Human CytokeRatin 8/KRT8 quantitates human Cytokeratin 8/KRT8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cytokeratin 8/KRT8.
Specifications
Specifications
| Accession Number | AA960620; AL022697; AU019895; C CY K2C8; CARD2; cb186; CK 8; CK8; CK-8; CYK8; CYKER; cytokeratin 8; cytokeratin 8 (370 AA); Cytokeratin endo A; cytokeratin-8; Cytokeratin-A; DreK8; EndoA; etID33550.23; etID43142.23; etID43142.8; hard keratin; K2C8; K8; K-8; KERA; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 8; keratin, type 2, gene 8; keratin, type II cytoskeletal 8; Keratin-8; KO; Krt-2.8; krt2-8; KRT8; LOW QUALITY PROTEIN: keratin, type II cytoskeletal 8; sb:cb186; type II cytokeratin A (no0.8); type-II keratin Kb8; wu:fa20h05; wu:fa95h10; wu:fb96c06; zfk8; zf-K8 |
| Assay Range | 0.410 to 100 pg/mL |
| Assay Sensitivity | 0.41 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 3856 |
| Gene Symbol | KRT8 |
| Interassay CV | <12% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.