Learn More
Biomatik Corporation Human Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12367
Encompass_Preferred
Human Cytokeratin Fragment Antigen 21-1 (CYFRA21-1) High-sensitive ELISA Kit for detecting CYFRA21-1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08727. Gene ID: 3880. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5120