Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 1 Kit.
Catalog No. 501948542
Encompass_Preferred
Detects 23 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: GCSF, GM-CSF, GRO alpha/beta/gamma, GRO alpha (CXCL1), IL-1 alpha (IL-1 F1), IL-2, IL-3, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-13, IL-15, IFN-gamma, MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MIG (CXCL9), RANTES (CCL5), TGF beta 1, TNF alpha, TNF beta (TNFSF1B)
Catalog No. 50-194-8542 Supplier Raybiotech Inc Supplier No. AAHCYT14
Detects 23 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: GCSF, GM-CSF, GRO alpha/beta/gamma, GRO alpha (CXCL1), IL-1 alpha (IL-1 F1), IL-2, IL-3, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-13, IL-15, IFN-gamma, MCP-1 (CCL2), MCP-2 (CCL8), MCP-3 (MARC/CCL7), MIG (CXCL9), RANTES (CCL5), TGF beta 1, TNF alpha, TNF beta (TNFSF1B)