Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 10 Kit.
Supplier:  Raybiotech Inc AAHCYT102
Encompass_Preferred
Detects 49 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 4-1BB (TNFRSF9/CD137), ACE-2, Alpha-fetoprotein, Angiopoietin-1, Angiostatin, ANGPTL4, Beta-2 Microglobulin, BCMA (TNFRSF17), beta IG-H3, CA125, CA15-3, CA19-9, CA9, Cathepsin S, HCC-1 (CCL14), 6Ckine (CCL21), CD23, CD40 Ligand (TNFSF5), CEA, CEACAM-1, Cripto-1, CRP (C-Reactive Protein), DAN, Decorin, DKK-1, Dkk-3, Dkk-4, CD26 (DPPIV), E-Cadherin, EDA-A2, EG-VEGF (PK1), TROP1 (EpCAM), ErbB2, Erythropoietin R, FSH, HB-EGF, hCG intact, HVEM (TNFRSF14), IL-13 R1, IL-17B, IL-17C, IL-17F, IL-17 RA, Procalcitonin, PSA-free, S100 B, Sonic Hedgehog N-Terminal (Shh-N), Thyroglobulin, Ubiquitin+1
Catalog No. 50-194-8532