Learn More
Raybiotech Inc HUMAN CYTOKINE ARRAY C1000 (4)
Catalog No. 501043084
Encompass_Preferred
RayBio C-Series Human Cytokine Antibody Array 1000 Kit. A combination of Human Cytokine Antibody Array C6 & C7. Detects 120 Human Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3084 Supplier Raybiotech Inc Supplier No. AAHCYT10004
Add to Cart
Add to Cart
RayBio C-Series Human Cytokine Antibody Array 1000 Kit. A combination of Human Cytokine Antibody Array C6 & C7. Detects 120 Human Cytokines. Suitable for all liquid sample types.