Learn More
Raybiotech Inc C-Series Human Receptor Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHCYT114
Encompass_Preferred
Detects 40 human cytokine receptor proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 2B4 (CD244), ADAM-9, APRIL, BMP-2, BMP-9, Cathepsin L, CD200, CD97, Chemerin, C5a, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGFR, IFN-alpha / beta R2, IGF-2 R, IL-1 R6 (IL-1 Rrp2), IL-24, IL-33 (IL-1 F11), Kallikrein 14, Legumain, LOX-1, MBL, Neprilysin, Notch-1, NOV (CCN3), Osteoactivin (GPNMB), PD-1, PGRPs, SERPINA4, sFRP-3, Thrombomodulin, TLR2, Transferrin, WIF-1
Catalog No. 50-194-8536