Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 12 Kit.
Catalog No. 501948540
Encompass_Preferred
Detects 32 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: aFGF (FGF-1), Albumin, AMICA, Angiopoietin-4, BAFF, CD163, Clusterin, CRTAM, CXCL14 (BRAK), Cystatin C, DLL1, Fetuin A, FOLR1, GASP-1 (WFIKKNRP), GASP-2 (WFIKKN), GCSF R (CD114), HAI-2, IL-17 RB, IL-27, LAG-3, LDL R, Pepsinogen 1, RBP4, SOST, Syndecan-1, TACI (TNFRSF13B), TFPI, Thrombospondin 1, TRANCE (TNFSF11), Troponin I, uPA, WISP-1 (CCN4)
Catalog No. 50-194-8540 Supplier Raybiotech Inc Supplier No. AAHCYT124
Detects 32 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: aFGF (FGF-1), Albumin, AMICA, Angiopoietin-4, BAFF, CD163, Clusterin, CRTAM, CXCL14 (BRAK), Cystatin C, DLL1, Fetuin A, FOLR1, GASP-1 (WFIKKNRP), GASP-2 (WFIKKN), GCSF R (CD114), HAI-2, IL-17 RB, IL-27, LAG-3, LDL R, Pepsinogen 1, RBP4, SOST, Syndecan-1, TACI (TNFRSF13B), TFPI, Thrombospondin 1, TRANCE (TNFSF11), Troponin I, uPA, WISP-1 (CCN4)