Learn More
Raybiotech Inc HUMAN CYTOKINE ARRAY C2000 (4)
Supplier:  Raybiotech Inc AAHCYT20004
Encompass_Preferred
RayBio C-Series Human Cytokine Antibody Array 2000 Kit. A combination of Human Cytokine Antibody Array C6, C7, & C8. Detects 174 Human Cytokines. Suitable for all liquid sample types.
Catalog No. 50-104-3099