Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 4 Kit.
Supplier:  Raybiotech Inc AAHCYT42
Encompass_Preferred
Detects 36 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BDNF, BLC (CXCL13), CK beta 8-1 (CCL23), Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), Eotaxin-3 (CCL26), FGF-4, FGF-6, FGF-7 (KGF), FGF-9, Flt-3 Ligand, Fractalkine (CX3CL1), GCP-2 (CXCL6), GDNF, HGF, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IL-16, LIF, LIGHT (TNFSF14), MCP-4 (CCL13), MIF, NAP-2 (PPBP/CXCL7), NT-3, NT-4, Osteoprotegerin (TNFRSF11B), PARC (CCL18), PLGF, TGF beta 2, TGF beta 3, TIMP-1, TIMP-2, MIP-3 alpha (CCL20), IP-10 (CXCL10)
Catalog No. 50-194-8553