Learn More
Raybiotech Inc C-Series Human Cytokine Antibody Array 4000 Kit.
Supplier:  Raybiotech Inc AAHCYT40002
Encompass_Preferred
A combination of Human Cytokine Antibody Array C6, C7, C8, C9, & C10. Detects 274 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAH-CYT-4000-2
Catalog No. 50-194-8550